Product Details

SNP ID
rs77302734
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:197215805 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGAGTCAATTAAACCTCCTTCC[C/T]GAATAAAGGACCCCAGAAGTTATTG
Phenotype
MIM: 604210
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRB1 PubMed Links

Gene Details

Gene
CRB1
Gene Name
crumbs 1, cell polarity complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193640.1 Intron NP_001180569.1
NM_001257965.1 Intron NP_001244894.1
NM_001257966.1 Intron NP_001244895.1
NM_201253.2 Intron NP_957705.1
XM_011509365.2 Intron XP_011507667.1
XM_011509367.1 Intron XP_011507669.1
XM_011509369.2 Intron XP_011507671.1
XM_017000851.1 Intron XP_016856340.1
XM_017000852.1 Intron XP_016856341.1

View Full Product Details