Product Details

SNP ID
rs80038126
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:155034073 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATCATCAGAATGGCACAAGAGGG[A/C]AAAGAAGAAAACAGCCTCATACCAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DCST1 PubMed Links

Gene Details

Gene
DCST1
Gene Name
DC-STAMP domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143687.2 151 Missense Mutation AAA,CAA K13Q NP_001137159.1
NM_152494.3 151 Missense Mutation AAA,CAA K13Q NP_689707.2
Gene
DCST2
Gene Name
DC-STAMP domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144622.2 151 Intron NP_653223.2
XM_011509188.2 151 Intron XP_011507490.1
XM_011509189.2 151 Intron XP_011507491.1

View Full Product Details