Product Details

SNP ID
rs77148982
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:155034080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAATGGCACAAGAGGGCAAAGAA[A/G]AAAACAGCCTCATACCAGTGAGTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCST1 PubMed Links

Gene Details

Gene
DCST1
Gene Name
DC-STAMP domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143687.2 158 Missense Mutation AAA,AGA K15R NP_001137159.1
NM_152494.3 158 Missense Mutation AAA,AGA K15R NP_689707.2
Gene
DCST2
Gene Name
DC-STAMP domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144622.2 158 Intron NP_653223.2
XM_011509188.2 158 Intron XP_011507490.1
XM_011509189.2 158 Intron XP_011507491.1

View Full Product Details