Product Details

SNP ID
rs78100219
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:156920674 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCGTAGGCTACGCCACCGCGGACG[A/G]GTCGGATCCGGTCCCTGGACGCGGA
Phenotype
MIM: 610278
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC71 PubMed Links

Gene Details

Gene
LRRC71
Gene Name
leucine rich repeat containing 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144702.2 43 UTR 5 NP_653303.2
XM_005244926.3 43 UTR 5 XP_005244983.1
XM_005244927.3 43 UTR 5 XP_005244984.1
XM_005244928.2 43 UTR 5 XP_005244985.1
XM_006711185.3 43 Intron XP_006711248.1
XM_006711186.3 43 UTR 5 XP_006711249.1
XM_006711187.3 43 UTR 5 XP_006711250.1
XM_011509239.2 43 Intron XP_011507541.1
XM_011509240.2 43 Intron XP_011507542.1
XM_011509241.2 43 UTR 5 XP_011507543.1
XM_017000459.1 43 UTR 5 XP_016855948.1
XM_017000460.1 43 Intron XP_016855949.1
XM_017000461.1 43 Intron XP_016855950.1
XM_017000462.1 43 Intron XP_016855951.1
Gene
PEAR1
Gene Name
platelet endothelial aggregation receptor 1
There are no transcripts associated with this gene.

View Full Product Details