Product Details

SNP ID
rs76007671
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:157577872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAATGGACCATCTGATGTAGAGTC[C/T]CCAGAAATGCATTGCTTTACTATCA
Phenotype
MIM: 605876
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FCRL4 PubMed Links

Gene Details

Gene
FCRL4
Gene Name
Fc receptor like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031282.2 Intron NP_112572.1
XM_011510034.1 Intron XP_011508336.1

View Full Product Details