Product Details

SNP ID
rs79989624
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:157578495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACCTTCCTCTTCTTCTCCCAGCT[G/T]AGTAGTCTGGATCTCAGAGTATACC
Phenotype
MIM: 605876
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FCRL4 PubMed Links

Gene Details

Gene
FCRL4
Gene Name
Fc receptor like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031282.2 1520 Missense Mutation AAG,CAG K470Q NP_112572.1
XM_011510034.1 1520 Missense Mutation AAG,CAG K469Q XP_011508336.1

View Full Product Details