Product Details
- SNP ID
-
rs79392937
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
14
- Location
-
Chr.1:158250289 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAATCAGTGCTGGGGAGGTGAAGTG[A/G]TGGGAGCCAAGACCACATGTCAGCA
- Phenotype
-
MIM: 188370
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CD1A
PubMed Links
Gene Details
- Gene
- CD1A
- Gene Name
- CD1a molecule
View Full Product Details