Product Details

SNP ID
rs79287493
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:158466039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTTCACTGTCTCCCTGGTCACC[A/C]CCTCCCTAGTATTTCATCTGCCCTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR10K1 PubMed Links

Gene Details

Gene
OR10K1
Gene Name
olfactory receptor family 10 subfamily K member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004473.1 478 Missense Mutation ACC,CCC T160P NP_001004473.1

View Full Product Details