Product Details

SNP ID
rs79211262
Assay Type
Functionally tested
NCBI dbSNP Submissions
27
Location
Chr.1:27976710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAAGTGCTAAGGCATTTTCCAAT[G/T]GGTATATACTTTGTTTGTTTTTTTT
Phenotype
MIM: 601655
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EYA3 PubMed Links

Gene Details

Gene
EYA3
Gene Name
EYA transcriptional coactivator and phosphatase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282560.1 Intron NP_001269489.1
NM_001282561.1 Intron NP_001269490.1
NM_001282562.1 Intron NP_001269491.1
NM_001990.3 Intron NP_001981.2
XM_006710449.3 Intron XP_006710512.1
XM_011540999.2 Intron XP_011539301.1
XM_011541001.2 Intron XP_011539303.1
XM_011541003.2 Intron XP_011539305.1
XM_011541004.2 Intron XP_011539306.1
XM_017000652.1 Intron XP_016856141.1
XM_017000653.1 Intron XP_016856142.1
XM_017000654.1 Intron XP_016856143.1
XM_017000655.1 Intron XP_016856144.1
XM_017000656.1 Intron XP_016856145.1
XM_017000657.1 Intron XP_016856146.1
XM_017000658.1 Intron XP_016856147.1
XM_017000659.1 Intron XP_016856148.1
Gene
XKR8
Gene Name
XK related 8
There are no transcripts associated with this gene.

View Full Product Details