Product Details

SNP ID
rs78423105
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:19266274 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACACGTCTTCAAGATGCACTTAC[A/C]TTGTAGATGTTTAACTACTTCTGTC
Phenotype
MIM: 608477 MIM: 608478
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AKR7A3 PubMed Links

Gene Details

Gene
AKR7A3
Gene Name
aldo-keto reductase family 7 member A3
There are no transcripts associated with this gene.

Gene
AKR7L
Gene Name
aldo-keto reductase family 7 like (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
MRTO4
Gene Name
MRT4 homolog, ribosome maturation factor
There are no transcripts associated with this gene.

View Full Product Details