Product Details

SNP ID
rs76383094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:164680582 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGGCCTGCTCACTTCTATTACT[C/T]ATATGGCAGATTCTGCTTCTTCCTT
Phenotype
MIM: 610318
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COBLL1 PubMed Links

Gene Details

Gene
COBLL1
Gene Name
cordon-bleu WH2 repeat protein like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278458.1 9087 UTR 3 NP_001265387.1
NM_001278460.1 9087 UTR 3 NP_001265389.1
NM_001278461.1 9087 UTR 3 NP_001265390.1
NM_014900.4 9087 UTR 3 NP_055715.3
Gene
SNORA70F
Gene Name
small nucleolar RNA, H/ACA box 70F
There are no transcripts associated with this gene.

View Full Product Details