Product Details

SNP ID
rs74502467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:171697415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGGGTGGGCCAATTGGTTTGTA[A/G]ACTGTCTCCCCTCCTGGGTTTGTCT
Phenotype
MIM: 603331
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DYNC1I2 PubMed Links

Gene Details

Gene
DYNC1I2
Gene Name
dynein cytoplasmic 1 intermediate chain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271785.1 Intron NP_001258714.1
NM_001271786.1 Intron NP_001258715.1
NM_001271787.1 Intron NP_001258716.1
NM_001271788.1 Intron NP_001258717.1
NM_001271789.1 Intron NP_001258718.1
NM_001271790.1 Intron NP_001258719.1
NM_001320882.1 Intron NP_001307811.1
NM_001320883.1 Intron NP_001307812.1
NM_001320884.1 Intron NP_001307813.1
NM_001378.2 Intron NP_001369.1
XM_006712347.3 Intron XP_006712410.1
XM_017003525.1 Intron XP_016859014.1

View Full Product Details