Product Details

SNP ID
rs79778627
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:182148334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTCATTTCTTGCTTGCTCTCCC[C/T]CCACTGGCTACTTTGGACTCCTAAG
Phenotype
MIM: 171890
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDE1A PubMed Links

Gene Details

Gene
PDE1A
Gene Name
phosphodiesterase 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003683.2 Intron NP_001003683.1
NM_001258312.1 Intron NP_001245241.1
NM_001258313.1 Intron NP_001245242.1
NM_001258314.1 Intron NP_001245243.1
NM_005019.4 Intron NP_005010.2
XM_011511323.2 Intron XP_011509625.1
XM_011511324.2 Intron XP_011509626.1
XM_011511325.2 Intron XP_011509627.1
XM_011511326.2 Intron XP_011509628.1
XM_017004294.1 Intron XP_016859783.1
XM_017004295.1 Intron XP_016859784.1
XM_017004296.1 Intron XP_016859785.1
XM_017004297.1 Intron XP_016859786.1
XM_017004298.1 Intron XP_016859787.1
XM_017004299.1 Intron XP_016859788.1
XM_017004300.1 Intron XP_016859789.1
XM_017004301.1 Intron XP_016859790.1
XM_017004302.1 Intron XP_016859791.1

View Full Product Details