Product Details

SNP ID
rs75674163
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:183079055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAACATGGAAAGATGCCAGAATTC[A/G]TGTTGTGGAAGAAGTAGAGCCGAGC
Phenotype
MIM: 611437
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUSP19 PubMed Links

Gene Details

Gene
DUSP19
Gene Name
dual specificity phosphatase 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142314.1 497 Missense Mutation CAT,CGT H41R NP_001135786.1
NM_001321519.1 497 Missense Mutation CAT,CGT H41R NP_001308448.1
NM_080876.3 497 Missense Mutation CAT,CGT H41R NP_543152.1

View Full Product Details