Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001195144.1 | Intron | NP_001182073.1 | |||
NM_153697.2 | Intron | NP_710181.2 | |||
XM_005246945.1 | Intron | XP_005247002.1 | |||
XM_005246946.1 | Intron | XP_005247003.1 | |||
XM_005246947.2 | Intron | XP_005247004.1 | |||
XM_005246948.2 | Intron | XP_005247005.1 | |||
XM_006712838.3 | Intron | XP_006712901.1 | |||
XM_011512148.2 | Intron | XP_011510450.1 | |||
XM_017005252.1 | Intron | XP_016860741.1 |