Product Details
- SNP ID
-
rs471839
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
30
- Location
-
Chr.1:35274196 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTATGCAATGTGAATAATTTGCTTT[A/T]AGAAAGTATTGACTCTTTGTTTAGA
- Phenotype
-
MIM: 613568
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ZMYM4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78994061] are located under a probe and SNP(s) [rs143443564] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZMYM4
- Gene Name
- zinc finger MYM-type containing 4
View Full Product Details