Product Details

SNP ID
rs159313
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:34357370 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGGGCTTGGGGGTGAGTAAGATCTA[C/T]TTCTTCCTCTTTGCTTTGCATCCCA
Phenotype
MIM: 182281 MIM: 601391
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCL1 PubMed Links

Gene Details

Gene
CCL1
Gene Name
C-C motif chemokine ligand 1
There are no transcripts associated with this gene.

Gene
CCL13
Gene Name
C-C motif chemokine ligand 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005408.2 Intron NP_005399.1

View Full Product Details