Product Details

SNP ID
rs621867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4340646 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCTGTTTCTAGCACCCCACCCA[C/T]GGGGGCTCCTGCTCCTTCGAAGTCC
Phenotype
MIM: 608539 MIM: 614336
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CORO7-PAM16 PubMed Links

Gene Details

Gene
CORO7-PAM16
Gene Name
CORO7-PAM16 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201479.1 Intron NP_001188408.1
Gene
GLIS2
Gene Name
GLIS family zinc finger 2
There are no transcripts associated with this gene.

Gene
PAM16
Gene Name
presequence translocase-associated motor 16 homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016069.9 Intron NP_057153.8

View Full Product Details