Product Details

SNP ID
rs454429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46468868 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTACCTCAATATGAGACATAATTA[C/T]TAATATGATCATTCTTTGGTATCAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MKRN7P PubMed Links

Gene Details

Gene
MKRN7P
Gene Name
makorin ring finger protein 7, pseudogene
There are no transcripts associated with this gene.

Gene
ZNF334
Gene Name
zinc finger protein 334
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270497.1 Intron NP_001257426.1
NM_018102.4 Intron NP_060572.3
XM_011528892.2 Intron XP_011527194.1
XM_011528895.2 Intron XP_011527197.1
XM_011528897.2 Intron XP_011527199.1
XM_017027933.1 Intron XP_016883422.1
XM_017027934.1 Intron XP_016883423.1
XM_017027935.1 Intron XP_016883424.1
XM_017027936.1 Intron XP_016883425.1
XM_017027937.1 Intron XP_016883426.1
XM_017027938.1 Intron XP_016883427.1
XM_017027939.1 Intron XP_016883428.1
XM_017027940.1 Intron XP_016883429.1
XM_017027941.1 Intron XP_016883430.1
XM_017027942.1 Intron XP_016883431.1
XM_017027943.1 Intron XP_016883432.1
XM_017027944.1 Intron XP_016883433.1

View Full Product Details