Product Details

SNP ID
rs282162
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:606311 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTGGCCTTCCTCTCTTTTACTCT[G/T]TAAGTGCTCAGGGACAGGGAGAGCC
Phenotype
MIM: 601010
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TCF15 PubMed Links

Gene Details

Gene
TCF15
Gene Name
transcription factor 15 (basic helix-loop-helix)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004609.3 Intron NP_004600.2

View Full Product Details