Product Details

SNP ID
rs396712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:61641067 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAGTACTGCTGACTGGACTAGTG[C/T]TCATCAAAGGTGGTCTGTGGACCAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C5orf64 PubMed Links

Gene Details

Gene
C5orf64
Gene Name
chromosome 5 open reading frame 64
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173667.3 Intron NP_775938.1
XM_017009378.1 Intron XP_016864867.1
XM_017009379.1 Intron XP_016864868.1
XM_017009380.1 Intron XP_016864869.1
XM_017009381.1 Intron XP_016864870.1
XM_017009382.1 Intron XP_016864871.1

View Full Product Details