Product Details

SNP ID
rs37630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:74466613 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGCTGTGTCCCTGGGCTGATTTCT[C/T]GTTGCAGGTCTCAGCTGGGTTGTCA
Phenotype
MIM: 604318
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GTF2IRD1 PubMed Links

Gene Details

Gene
GTF2IRD1
Gene Name
GTF2I repeat domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199207.1 Intron NP_001186136.1
NM_005685.3 Intron NP_005676.3
NM_016328.2 Intron NP_057412.1
XM_006716182.3 Intron XP_006716245.1
XM_006716183.3 Intron XP_006716246.1
XM_011516713.1 Intron XP_011515015.1
XM_017012802.1 Intron XP_016868291.1
XM_017012803.1 Intron XP_016868292.1
XM_017012804.1 Intron XP_016868293.1
XM_017012805.1 Intron XP_016868294.1

View Full Product Details