Product Details

SNP ID
rs224496
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3610734 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCATCACCCTCTCCACGCCCCA[C/G]TCCTGGAGCTGCTGAATCGGAAGCA
Phenotype
MIM: 605060 MIM: 602076
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SHPK PubMed Links

Gene Details

Gene
SHPK
Gene Name
sedoheptulokinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013276.2 1366 Missense Mutation GAC,GAG D421E NP_037408.2
Gene
TRPV1
Gene Name
transient receptor potential cation channel subfamily V member 1
There are no transcripts associated with this gene.

View Full Product Details