Product Details

SNP ID
rs34511020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58409929 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGGTTGTTTTTTTCTGCCCATC[A/G]TTGACCCCAAGGCTCAGTTGGACCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100506634 PubMed Links

Gene Details

Gene
LOC100506634
Gene Name
hepatocellular carcinoma-associated antigen HCA25a
There are no transcripts associated with this gene.

Gene
ZNF584
Gene Name
zinc finger protein 584
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318002.1 Intron NP_001304931.1
NM_173548.2 Intron NP_775819.1
XM_005258628.4 Intron XP_005258685.1
XM_005258630.4 Intron XP_005258687.1

View Full Product Details