Product Details

SNP ID
rs2217988
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:51322866 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTACACTTGGGCAGTGGCATCCA[C/T]CCATTCATTACTCAATAGATATATT
Phenotype
MIM: 615904
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PXDNL PubMed Links

Gene Details

Gene
PXDNL
Gene Name
peroxidasin like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144651.4 Intron NP_653252.3
XM_005251168.3 Intron XP_005251225.1
XM_006716420.3 Intron XP_006716483.1
XM_011517456.2 Intron XP_011515758.1
XM_011517457.2 Intron XP_011515759.1
XM_011517458.2 Intron XP_011515760.1
XM_011517459.2 Intron XP_011515761.1
XM_017013041.1 Intron XP_016868530.1

View Full Product Details