Product Details

SNP ID
rs1052489
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:6723861 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTACTGTATGATGTTTTATTTAG[C/T]GAACAGTTCACATACCCAAATGGTT
Phenotype
MIM: 605469
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KDM4C PubMed Links

Gene Details

Gene
KDM4C
Gene Name
lysine demethylase 4C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146695.1 Intron NP_001140167.1
NM_001146696.1 Intron NP_001140168.1
NM_001304339.1 Intron NP_001291268.1
NM_001304340.1 Intron NP_001291269.1
NM_001304341.1 Intron NP_001291270.1
NM_015061.3 Intron NP_055876.2
XM_006716741.2 Intron XP_006716804.1
XM_011517811.2 Intron XP_011516113.1
XM_011517812.2 Intron XP_011516114.2
XM_011517816.2 Intron XP_011516118.1
XM_017014498.1 Intron XP_016869987.1
XM_017014499.1 Intron XP_016869988.1
XM_017014500.1 Intron XP_016869989.1
XM_017014501.1 Intron XP_016869990.1
XM_017014502.1 Intron XP_016869991.1
XM_017014503.1 Intron XP_016869992.1
XM_017014504.1 Intron XP_016869993.1
XM_017014505.1 Intron XP_016869994.1
XM_017014506.1 Intron XP_016869995.1

View Full Product Details