Product Details

SNP ID
rs35015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:62391714 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGATACGCATGAGGCTCACAACCAG[A/G]AATCTGCTATTGAGCCATATGAACT
Phenotype
MIM: 612499 MIM: 602591
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DIMT1 PubMed Links

Gene Details

Gene
DIMT1
Gene Name
DIM1 dimethyladenosine transferase 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014473.2 Intron NP_055288.1
XM_011543323.2 Intron XP_011541625.1
Gene
KIF2A
Gene Name
kinesin family member 2A
There are no transcripts associated with this gene.

View Full Product Details