Product Details
- SNP ID
-
rs13156073
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:62395530 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TATTTCAAAACAAAAGTTTCATGAA[A/G]CCATAATCACCTGGACTACATGCAA
- Phenotype
-
MIM: 612499
MIM: 602591
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DIMT1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74333974] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DIMT1
- Gene Name
- DIM1 dimethyladenosine transferase 1 homolog
- Gene
- KIF2A
- Gene Name
- kinesin family member 2A
There are no transcripts associated with this gene.
View Full Product Details