Product Details

SNP ID
rs828891
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:74162821 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACTCTTAAGTTGAAAATGACAAGGA[G/T]TGATTTCATCCATGAATACTGATGC
Phenotype
MIM: 609281
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MOB1A PubMed Links

Gene Details

Gene
MOB1A
Gene Name
MOB kinase activator 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317110.1 Intron NP_001304039.1
NM_001317111.1 Intron NP_001304040.1
NM_001317112.1 Intron NP_001304041.1
NM_018221.4 Intron NP_060691.2
XM_011532948.2 Intron XP_011531250.1

View Full Product Details