Product Details

SNP ID
rs34153334
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:36534973 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGGGGGTCGGCCGGCTCCTCCCTA[G/T]GGGCGGGGCGAGCACTCGCGGCGCG
Phenotype
MIM: 612275 MIM: 610275
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GGNBP2 PubMed Links

Gene Details

Gene
GGNBP2
Gene Name
gametogenetin binding protein 2
There are no transcripts associated with this gene.

Gene
MYO19
Gene Name
myosin XIX
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033580.2 485 UTR 5 NP_001028752.1
NM_001163735.1 485 UTR 5 NP_001157207.1
NM_025109.5 485 UTR 5 NP_079385.2
XM_011525286.1 485 Intron XP_011523588.1
XM_011525287.1 485 Intron XP_011523589.1
XM_011525290.2 485 Intron XP_011523592.1
XM_017025157.1 485 Intron XP_016880646.1
XM_017025158.1 485 Intron XP_016880647.1
XM_017025159.1 485 Intron XP_016880648.1
XM_017025160.1 485 Intron XP_016880649.1
XM_017025161.1 485 Intron XP_016880650.1
Gene
PIGW
Gene Name
phosphatidylinositol glycan anchor biosynthesis class W
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178517.3 485 Intron NP_848612.2
XM_005257238.2 485 UTR 5 XP_005257295.1
XM_011524646.2 485 Intron XP_011522948.1

View Full Product Details