Product Details
- SNP ID
-
rs2668760
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:47156520 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GTGCAACCCCAAGAACATGAGTGCA[C/T]TGGAGCCTGGCAGACGTAGACCCAC
- Phenotype
-
MIM: 609382
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HDHD2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12326221] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HDHD2
- Gene Name
- haloacid dehalogenase like hydrolase domain containing 2
There are no transcripts associated with this gene.
- Gene
- IER3IP1
- Gene Name
- immediate early response 3 interacting protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016097.4 |
|
Intron |
|
|
NP_057181.1 |
View Full Product Details