Product Details

SNP ID
rs2932775
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:69224458 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTATAGTGTTTTCTTCTTTGTGCT[C/T]TTCTGTGTTCTTCCAAATGTTTTAC
Phenotype
MIM: 611996
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPS36 PubMed Links

Gene Details

Gene
MRPS36
Gene Name
mitochondrial ribosomal protein S36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033281.5 Intron NP_150597.1

View Full Product Details