Product Details
- SNP ID
-
rs11590355
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
22
- Location
-
Chr.1:108888029 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTAAATATTATCATCATTTAATAA[A/G]GTGATTAGGGAATATGTTTTTGTTT
- Phenotype
-
MIM: 609245
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GPSM2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs150392894] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GPSM2
- Gene Name
- G-protein signaling modulator 2
View Full Product Details