Product Details

SNP ID
rs11676396
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:237803532 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTATGTAATCTGAATAAGCGTCT[C/T]CTTTTGGATATATGGATTGTGAATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBM44 PubMed Links

Gene Details

Gene
RBM44
Gene Name
RNA binding motif protein 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080504.2 Intron NP_001073973.2
XM_005246075.1 Intron XP_005246132.1
XM_011511152.2 Intron XP_011509454.1
XM_011511153.2 Intron XP_011509455.1
XM_011511154.2 Intron XP_011509456.1
XM_011511155.1 Intron XP_011509457.1
XM_017004054.1 Intron XP_016859543.1
XM_017004055.1 Intron XP_016859544.1
XM_017004056.1 Intron XP_016859545.1
XM_017004057.1 Intron XP_016859546.1
XM_017004058.1 Intron XP_016859547.1

View Full Product Details