Product Details

SNP ID
rs1550637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:90956116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGAAACGCAGTTGGACTCAGGTG[A/G]ATTGGCTGCGGTCCCAAGGCCCCGC
Phenotype
MIM: 603484 MIM: 611219
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRC1 PubMed Links

Gene Details

Gene
PRC1
Gene Name
protein regulator of cytokinesis 1
There are no transcripts associated with this gene.

Gene
RCCD1
Gene Name
RCC1 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017919.1 Intron NP_001017919.1
NM_033544.2 Intron NP_291022.2
Gene
UNC45A
Gene Name
unc-45 myosin chaperone A
There are no transcripts associated with this gene.

View Full Product Details