Product Details

SNP ID
rs7177338
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:90885406 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCCTGCCTCCCCCATCTGTGCT[A/G]TATAGTCCTGGGCTGAGATCACCAG
Phenotype
MIM: 190030 MIM: 136950
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FES PubMed Links

Gene Details

Gene
FES
Gene Name
FES proto-oncogene, tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143783.1 Intron NP_001137255.1
NM_001143784.1 Intron NP_001137256.1
NM_001143785.1 Intron NP_001137257.1
NM_002005.3 Intron NP_001996.1
XM_005254880.1 Intron XP_005254937.1
XM_005254882.1 Intron XP_005254939.1
XM_017022005.1 Intron XP_016877494.1
XM_017022006.1 Intron XP_016877495.1
XM_017022007.1 Intron XP_016877496.1
XM_017022008.1 Intron XP_016877497.1
XM_017022009.1 Intron XP_016877498.1
XM_017022010.1 Intron XP_016877499.1
Gene
FURIN
Gene Name
furin, paired basic amino acid cleaving enzyme
There are no transcripts associated with this gene.

View Full Product Details