Product Details

SNP ID
rs6413436
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:913513 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATTTTAAAATAAACTGACAGCTA[A/G]TGATTTCTGGATTATATTAATGATC
Phenotype
MIM: 600392 MIM: 605232
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
RAD52 PubMed Links

Gene Details

Gene
RAD52
Gene Name
RAD52 homolog, DNA repair protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297419.1 Intron NP_001284348.1
NM_001297420.1 Intron NP_001284349.1
NM_001297421.1 Intron NP_001284350.1
NM_001297422.1 Intron NP_001284351.1
NM_134424.3 Intron NP_602296.2
XM_005253720.4 Intron XP_005253777.1
XM_005253721.2 Intron XP_005253778.1
XM_011520990.2 Intron XP_011519292.1
XM_011520991.2 Intron XP_011519293.1
XM_011520993.2 Intron XP_011519295.1
XM_011520995.2 Intron XP_011519297.1
XM_017019769.1 Intron XP_016875258.1
XM_017019770.1 Intron XP_016875259.1
XM_017019771.1 Intron XP_016875260.1
Gene
WNK1
Gene Name
WNK lysine deficient protein kinase 1
There are no transcripts associated with this gene.

View Full Product Details