Product Details

SNP ID
rs11825335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:70405552 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGTTGCATATGACTTAGAGGAGA[C/G]AATTGCAGAAAACTCTGGAATTGAG
Phenotype
MIM: 164765
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CTTN PubMed Links

Gene Details

Gene
CTTN
Gene Name
cortactin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184740.1 Intron NP_001171669.1
NM_005231.3 Intron NP_005222.2
NM_138565.2 Intron NP_612632.1
XM_006718447.2 Intron XP_006718510.1
XM_006718448.3 Intron XP_006718511.1
XM_017017312.1 Intron XP_016872801.1

View Full Product Details