Product Details

SNP ID
rs12805811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67405769 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCAGGTGCTCCAGCCCACTTTCCC[C/T]AGCCCAGCTCTACAGTCTTGCATCT
Phenotype
MIM: 613368 MIM: 176875 MIM: 603761 MIM: 610831
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CARNS1 PubMed Links

Gene Details

Gene
CARNS1
Gene Name
carnosine synthase 1
There are no transcripts associated with this gene.

Gene
PPP1CA
Gene Name
protein phosphatase 1 catalytic subunit alpha
There are no transcripts associated with this gene.

Gene
RAD9A
Gene Name
RAD9 checkpoint clamp component A
There are no transcripts associated with this gene.

Gene
TBC1D10C
Gene Name
TBC1 domain family member 10C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256508.1 Intron NP_001243437.1
NM_198517.3 Intron NP_940919.1
XM_006718538.3 Intron XP_006718601.1
XM_006718539.3 Intron XP_006718602.1
XM_006718540.3 Intron XP_006718603.1
XM_006718541.3 Intron XP_006718604.1
XM_006718542.3 Intron XP_006718605.1
XM_006718543.2 Intron XP_006718606.1
XM_006718544.3 Intron XP_006718607.1
XM_011545001.2 Intron XP_011543303.1
XM_011545002.2 Intron XP_011543304.1

View Full Product Details