Product Details

SNP ID
rs35674448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102133366 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGGGCCGGAGCAAGTGGGCCCTA[A/G]GCCGTTTGCTGGGGGTAGGCGGGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPRC1 PubMed Links

Gene Details

Gene
PPRC1
Gene Name
peroxisome proliferator-activated receptor gamma, coactivator-related 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288727.1 Intron NP_001275656.1
NM_001288728.1 Intron NP_001275657.1
NM_015062.4 Intron NP_055877.3
XM_005269656.3 Intron XP_005269713.1
XM_006717731.3 Intron XP_006717794.1
XM_011539549.2 Intron XP_011537851.1
XM_011539551.2 Intron XP_011537853.1
XM_011539552.2 Intron XP_011537854.1
XM_017015982.1 Intron XP_016871471.1
XM_017015983.1 Intron XP_016871472.1
XM_017015984.1 Intron XP_016871473.1
XM_017015985.1 Intron XP_016871474.1

View Full Product Details