Product Details

SNP ID
rs12541569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:109552947 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCATCTCTATGGAAAGGAAAAACA[C/G]GCTTGTATCATAAATTTCATAAAGG
Phenotype
MIM: 605772
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EBAG9 PubMed Links

Gene Details

Gene
EBAG9
Gene Name
estrogen receptor binding site associated, antigen, 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278938.1 Intron NP_001265867.1
NM_004215.4 Intron NP_004206.1
NM_198120.2 Intron NP_936056.1
XM_017013960.1 Intron XP_016869449.1

View Full Product Details