Product Details
- SNP ID
-
rs870424
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:74033328 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGAAGGGAGAGAGGCACGTGAAGAA[C/T]GAGGGAGAGGGGGAGAAAGAGGAGG
- Phenotype
-
MIM: 130160
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ELN
PubMed Links
Gene Details
- Gene
- ELN
- Gene Name
- elastin
View Full Product Details