Product Details

SNP ID
rs1180272
Assay Type
Validated
NCBI dbSNP Submissions
40
Location
Chr.1:39659725 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GATAACTATCAGGGCAACTGTTGAT[A/G]TGTATCATCCATCCATATGCCCATC
Phenotype
MIM: 610525
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NT5C1A PubMed Links

Gene Details

Gene
NT5C1A
Gene Name
5'-nucleotidase, cytosolic IA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032526.2 Intron NP_115915.1

View Full Product Details