Product Details

SNP ID
rs1710146
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:120509403 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAGCTCCATCAGATTAAGAGCTGAA[C/T]GTTCCCCTTTTCCTGTAGAAAAGCC
Phenotype
MIM: 616359
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COQ5 PubMed Links

Gene Details

Gene
COQ5
Gene Name
coenzyme Q5, methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032314.3 Intron NP_115690.3
XM_006719639.2 Intron XP_006719702.1

View Full Product Details