Product Details

SNP ID
rs2295362
Assay Type
Validated
NCBI dbSNP Submissions
40
Location
Chr.1:1923880 on Build GRCh38
Set Membership
JSNP Validated
Context Sequence [VIC/FAM]
GTGTCTTCCCGGGGTCCATGACGCC[T/C]TTGACGGGGGCCACGCTGAACACGC
Phenotype
MIM: 610171
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
CALML6 PubMed Links

Gene Details

Gene
CALML6
Gene Name
calmodulin like 6
There are no transcripts associated with this gene.

Gene
CFAP74
Gene Name
cilia and flagella associated protein 74
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304360.1 3869 Silent Mutation AAA,AAG K1428K NP_001291289.1
XM_011542333.2 3869 Silent Mutation AAA,AAG K1451K XP_011540635.2
XM_011542335.1 3869 Silent Mutation AAA,AAG K1259K XP_011540637.1
XM_011542336.2 3869 Silent Mutation AAA,AAG K1259K XP_011540638.1
XM_017002641.1 3869 Silent Mutation AAA,AAG K1470K XP_016858130.1
XM_017002642.1 3869 Silent Mutation AAA,AAG K1470K XP_016858131.1
XM_017002643.1 3869 Silent Mutation AAA,AAG K1470K XP_016858132.1
XM_017002644.1 3869 Silent Mutation AAA,AAG K614K XP_016858133.1
Gene
TMEM52
Gene Name
transmembrane protein 52
There are no transcripts associated with this gene.

View Full Product Details