Product Details

SNP ID
rs12407630
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:87336496 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCATTGATAGTAAACTAAAAATA[C/T]GATTTGCCCTCCATCTCTTAATGCC
Phenotype
MIM: 603129
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LMO4 PubMed Links

Gene Details

Gene
LMO4
Gene Name
LIM domain only 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006769.3 Intron NP_006760.1
XM_005271291.3 Intron XP_005271348.1

View Full Product Details