Product Details

SNP ID
rs11582178
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:67142417 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAGCCATCCCATTTTAAATGCTT[C/T]TCTACTCTTTATTCCAGTTTTGTGG
Phenotype
MIM: 607562
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf141 PubMed Links

Gene Details

Gene
C1orf141
Gene Name
chromosome 1 open reading frame 141
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276351.1 Intron NP_001263280.1
NM_001276352.1 Intron NP_001263281.1
XM_011541465.2 Intron XP_011539767.1
XM_011541466.2 Intron XP_011539768.1
XM_011541467.1 Intron XP_011539769.1
XM_011541469.1 Intron XP_011539771.1
XM_011541472.1 Intron XP_011539774.1
XM_011541473.2 Intron XP_011539775.1
XM_011541475.1 Intron XP_011539777.1
XM_017001276.1 Intron XP_016856765.1
XM_017001277.1 Intron XP_016856766.1
Gene
IL23R
Gene Name
interleukin 23 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144701.2 Intron NP_653302.2
XM_005270516.2 Intron XP_005270573.1
XM_011540790.2 Intron XP_011539092.1
XM_011540791.2 Intron XP_011539093.1

View Full Product Details