Product Details
- SNP ID
-
rs12614187
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:181537764 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAAAAAAAAGAATTTGAATTGATAT[C/T]TAAAAACAGAATTTGAATTGATATT
- Phenotype
-
MIM: 608381
MIM: 192975
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CERKL
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74486071] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CERKL
- Gene Name
- ceramide kinase like
- Gene
- ITGA4
- Gene Name
- integrin subunit alpha 4
View Full Product Details