Product Details

SNP ID
rs6717512
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:186702911 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCAACTTCTATTTAATATGCTTTT[G/T]CTTCCTCAAGCAGATTTCTATATCA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM171B PubMed Links

Gene Details

Gene
FAM171B
Gene Name
family with sequence similarity 171 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_177454.3 Intron NP_803237.3

View Full Product Details