Product Details

SNP ID
rs35273375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:10112357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGACTGCATGTAGTTATGGGCT[A/G]CAGCTCTATCTGCATCACCCAGGAA
Phenotype
MIM: 138253
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRIN2A PubMed Links

Gene Details

Gene
GRIN2A
Gene Name
glutamate ionotropic receptor NMDA type subunit 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000833.4 Intron NP_000824.1
NM_001134407.2 Intron NP_001127879.1
NM_001134408.2 Intron NP_001127880.1
XM_011522458.2 Intron XP_011520760.1
XM_011522461.2 Intron XP_011520763.1
XM_017023172.1 Intron XP_016878661.1
XM_017023173.1 Intron XP_016878662.1
Gene
IMPDH1P11
Gene Name
inosine monophosphate dehydrogenase 1 pseudogene 11
There are no transcripts associated with this gene.

View Full Product Details